2-Methyl-2H-pyrazole-3-sulfonyl chloride

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1H-pyrazole-5-sulphonyl chloride

054453-500MG
101907-372EA 300 USD
101907-372
2-Methyl-2H-pyrazole-3-sulfonyl chloride
2-Methyl-2H-pyrazole-3-sulfonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₄H₅ClN₂O₂S
MW: 180.61 g/mol
MDL Number: MFCD09065021
CAS Number: 1020721-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR