tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl N-[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]carbamate

054332-500MG 054332-1G
101907-090EA 453.85 USD
101907-090 101907-092
tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate ≥95%
tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate

Hazard: Irritant

Formula: C₉H₁₉NO₅
MW: 221.26 g/mol
MDL Number: MFCD14581676
CAS Number: 146651-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR