5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one Hydrochloride

054267-1G 054267-5G
101906-832EA 169.23 USD
101906-832 101906-834
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥95%
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNOS
MDL Number: MFCD11111130
CAS Number: 115473-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR