Fmoc-cis-4-fluoro-Pro-OH

Supplier: Matrix Scientific

042060-500MG 042060-1G
101854-288EA 598.46 USD
101854-288 101854-290
Fmoc-cis-4-fluoro-Pro-OH
Fmoc-cis-4-fluoro-Pro-OH

Hazard: Irritant

Formula: C₂₀H₁₈FNO₄
MDL Number: MFCD07781261
CAS Number: 203866-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR