trans-N-Boc-4-cyano-L-proline methyl ester
Supplier: Matrix Scientific
Synonyms:
1-tert-Butyl 2-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate
042036-1G
042036-500MG
101854-192EA
1207.69
USD
101854-192
101854-194
trans-N-Boc-4-cyano-L-proline methyl ester
trans-N-Boc-4-cyano-L-proline methyl ester
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₄ MW: 254.286 g/mol |
MDL Number:
MFCD03094915 CAS Number: 194163-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...