Fmoc-3-(trifluoromethyl)-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3-CF₃)-OHFmoc-Phe(3-CF3)-OH

041932-1G 041932-5G
101853-784EA 4004.62 USD
101853-784 101853-782
Fmoc-3-(trifluoromethyl)-L-phenylalanine
Fmoc-3-(trifluoromethyl)-L-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₀F₃NO₄
MW: 455.44 g/mol
MDL Number: MFCD00672555
CAS Number: 205526-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR