Boc-S-trityl-L-homocysteine
Supplier: Matrix Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-4-(tritylthio)butanoic acid, Boc-Homocys(Trt)-OH, Boc-HoCys(Trt)-OH
041775-1G
041775-5G
101853-138EA
661.54
USD
101853-138
101853-140
Boc-S-trityl-L-homocysteine
Boc-S-trityl-L-homocysteine
CAS: 201419-16-1
MDL Number: MFCD00672309
Molecular Formula: C28H31NO4S
Molecular Weight: 477,63
Hazard: Irritant
Formula:
C₂₈H₃₁NO₄S MW: 477.619 g/mol |
MDL Number:
MFCD00672309 CAS Number: 201419-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...