Boc-D-3-Pyridylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-β-(3-pyridyl)-D-Ala-OHBoc-D-3-Pal-OHBoc-3-(3-pyridyl)-D-Ala-OH

041433-1G 041433-5G
101851-760EA 113.85 USD
101851-760 101851-758
Boc-D-3-Pyridylalanine ≥95%
Boc-D-3-Pyridylalanine

Purity: 95%
CAS: 98266-33-2
MDL Number: MFCD00076901
Molecular Formula: C13H18N2O4
Molecular Weight: 266,3
Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
MDL Number: MFCD00076901
CAS Number: 98266-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR