4-Amino-N-methylbenzamide ≥97%

Supplier: Matrix Scientific

025850-1G 025850-5G
101821-102EA 15.38 USD
101821-102 101821-104
4-Amino-N-methylbenzamide ≥97%
4-Amino-N-methylbenzamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MDL Number: MFCD00665792
CAS Number: 6274-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR