Thallium(I) hexafluorophosphate ≥98%

Supplier: Matrix Scientific

Synonyms: Thallium(I) hexafluorophosphate(V)

009174-5G
101796-566EA 270.77 USD
101796-566
Thallium(I) hexafluorophosphate ≥98%
Thallium (I) hexafluorophosphate

Hazard: Toxic

Formula: F₆PTl
MDL Number: MFCD00049807
CAS Number: 60969-19-9
UN: 1707
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR