1,4-Dichlorohexafluoro-2-butene ≥97%

Supplier: Matrix Scientific

008900-5G 008900-25G
101795-760EA 438.46 USD
101795-760 101795-762
1,4-Dichlorohexafluoro-2-butene ≥97%
1,4-Dichlorohexafluoro-2-butene

Hazard: Irritant

Formula: C₄Cl₂F₆
Boiling Pt: 64…66 °C
MDL Number: MFCD00236665
CAS Number: 360-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR