4-Ethoxynicotinaldehyde ≥95%

Supplier: Matrix Scientific

053826-500MG 053826-2.5G
101905-156EA 98.46 USD
101905-156 101905-158
4-Ethoxynicotinaldehyde ≥95%
4-Ethoxynicotinaldehyde

Hazard: Irritant

Formula: C₈H₉NO₂
MDL Number: MFCD13562930
CAS Number: 1220021-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR