4,5,6,7-Tetrahydro-1-benzothiophen-4-amine

Supplier: Matrix Scientific

053797-500MG 053797-2.5G
101905-044EA 364.62 USD
101905-044 101905-046
4,5,6,7-Tetrahydro-1-benzothiophen-4-amine
4,5,6,7-Tetrahydro-1-benzothiophen-4-amine

Hazard: Irritant

Formula: C₈H₁₁NS
MDL Number: MFCD13192826
CAS Number: 58094-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR