N-(3-Ethoxypropyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053296-500MG
053296-2.5G
101903-070EA
364.62
USD
101903-070
101903-072
N-(3-Ethoxypropyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Ethoxypropyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD13562696 CAS Number: 1236257-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...