3-Amino-N-(tert-butyl)-4-chlorobenzenesulfonamide

Supplier: Matrix Scientific

052911-500MG 052911-2.5G
101901-532EA 364.62 USD
101901-532 101901-534
3-Amino-N-(tert-butyl)-4-chlorobenzenesulfonamide
3-Amino-N-(tert-butyl)-4-chlorobenzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O₂S
MDL Number: MFCD09900856
CAS Number: 1017459-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR