Ethyl-3-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049517-500MG 049517-2.5G
101887-972EA 364.62 USD
101887-972 101887-974
Ethyl-3-(3-azetidinyloxy)benzoate
Ethyl-3-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD09861906
CAS Number: 954223-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR