3-(2-Chloro-4,6-dimethylphenoxy)azetidine
Supplier: Matrix Scientific
049415-500MG
049415-2.5G
101887-564EA
364.62
USD
101887-564
101887-566
3-(2-Chloro-4,6-dimethylphenoxy)azetidine
3-(2-Chloro-4,6-dimethylphenoxy)azetidine
Hazard: Irritant
Formula:
C₁₁H₁₄ClNO |
MDL Number:
MFCD13559884 CAS Number: 1219960-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...