1-Isobutyryl-3-piperidinecarboxylic acid
Supplier: Matrix Scientific
049272-500MG
049272-2.5G
101886-992EA
186.15
USD
101886-992
101886-994
1-Isobutyryl-3-piperidinecarboxylic acid
1-Isobutyryl-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₇NO₃ |
MDL Number:
MFCD09050645 CAS Number: 926214-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...