N-Isobutyrylalanine

Supplier: Matrix Scientific

049269-500MG 049269-2.5G
101886-980EA 364.62 USD
101886-980 101886-982
N-Isobutyrylalanine
N-Isobutyrylalanine

Hazard: Irritant

Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
MDL Number: MFCD09046347
CAS Number: 157170-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR