2-Methyl-N-propionylalanine
Supplier: Matrix Scientific
049253-500MG
049253-2.5G
101886-916EA
364.62
USD
101886-916
101886-918
2-Methyl-N-propionylalanine
2-Methyl-N-propionylalanine
Hazard: Irritant
Formula:
C₇H₁₃NO₃ MW: 159.19 g/mol |
MDL Number:
MFCD13559776 CAS Number: 106897-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...