1-Propionyl-4-piperidinecarboxylic acid
Supplier: Matrix Scientific
049252-500MG
049252-2.5G
101886-912EA
364.62
USD
101886-912
101886-914
1-Propionyl-4-piperidinecarboxylic acid
1-Propionyl-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₉H₁₅NO₃ |
MDL Number:
MFCD01414449 CAS Number: 117705-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...