Methyl (2S,4S)-4-(propionyloxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049248-500MG 049248-2.5G
101886-896EA 364.62 USD
101886-896 101886-898
Methyl (2S,4S)-4-(propionyloxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(propionyloxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₄
MDL Number: MFCD13559775
CAS Number: 1354487-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR