2-(3-Piperidinyl)ethyl propanoate hydrochloride
Supplier: Matrix Scientific
049244-500MG
049244-2.5G
101886-880EA
364.62
USD
101886-880
101886-882
2-(3-Piperidinyl)ethyl propanoate hydrochloride
2-(3-Piperidinyl)ethyl propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₂ MW: 221.73 g/mol |
MDL Number:
MFCD13559771 CAS Number: 1220037-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...