3-Azetidinyl propionate
Supplier: Matrix Scientific
049240-500MG
049240-2.5G
101886-864EA
364.62
USD
101886-864
101886-866
3-Azetidinyl propionate
3-Azetidinyl propionate
Hazard: Irritant
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol |
MDL Number:
MFCD13559767 CAS Number: 1220037-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...