2-(3-Piperidinyl)ethyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049142-500MG 049142-2.5G
101886-472EA 364.62 USD
101886-472 101886-474
2-(3-Piperidinyl)ethyl-2-chloroacetate hydrochloride
2-(3-Piperidinyl)ethyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₇Cl₂NO₂
MDL Number: MFCD13559698
CAS Number: 1220020-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR