N-(2-Methoxyacetyl)-2-methylalanine

Supplier: Matrix Scientific

049133-500MG 049133-2.5G
101886-436EA 364.62 USD
101886-436 101886-438
N-(2-Methoxyacetyl)-2-methylalanine
N-(2-Methoxyacetyl)-2-methylalanine

Hazard: Irritant

Formula: C₇H₁₃NO₄
MW: 175.19 g/mol
MDL Number: MFCD13559691
CAS Number: 1220037-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR