5-Fluoro-2-iodotoluene ≥98%

Supplier: Matrix Scientific

002140-25G 002140-100G
101783-310EA 32.31 USD
101783-310 101783-312
5-Fluoro-2-iodotoluene ≥98%
5-Fluoro-2-iodotoluene

Hazard: Irritant

Formula: C₇H₆FI
Boiling Pt: 98 °C (12 Torr)
MDL Number: MFCD00039397
CAS Number: 66256-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR