tert-Butyl (piperazin-1-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-(piperazin-1-yl)acetate, 1-[(tert-Butoxycarbonyl)methyl]piperazine, (Piperazin-1-yl)acetic acid tert-butyl ester

043268-500MG 043268-1G 043268-5G
101859-984EA 209.23 USD
101859-984 101859-986 101859-988
tert-Butyl (piperazin-1-yl)acetate ≥95%
tert-Butyl (piperazin-1-yl)acetate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 76 °C (0.2 mmHg)
MDL Number: MFCD06659535
CAS Number: 112257-22-4

Order Now

Specification Test Results

Boiling point 76°C/0.2mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR