3-(4'-Methoxyphenyl)propyl chloride ≥97%
Supplier: Matrix Scientific
007617-1G
007617-5G
101793-124EA
915.38
USD
101793-124
101793-126
3-(4'-Methoxyphenyl)propyl chloride ≥97%
3-(4'-Methoxyphenyl)propyl chloride
Hazard: Irritant
Formula:
C₁₀H₁₃ClO Boiling Pt: 100 °C ( .3 mmHg) |
MDL Number:
MFCD00045303 CAS Number: 59623-12-0 |
Specification Test Results
Boiling point | 100°C/ .3mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...