5-Iodooctafluoro-3-oxapentanesulfonyl fluoride ≥95%
Supplier: Matrix Scientific
002942-1G
002942-5G
101784-950EA
140
USD
101784-950
101784-952
5-Iodooctafluoro-3-oxapentanesulfonyl fluoride ≥95%
5-Iodooctafluoro-3-oxapentanesulfonyl fluoride
Purity: 95%
CAS: 66137-74-4
MDL Number: MFCD00798139
Molecular Formula: C4F9IO3S
Molecular Weight: 426
Boiling Point: 126-127°C
Hazard: Irritant
Formula:
C₄F₉IO₃S Boiling Pt: 126…127 °C Flash Pt: >100 °C |
MDL Number:
MFCD00798139 CAS Number: 66137-74-4 |
Specification Test Results
Boiling point | 126°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...