1H,1H,2H,3H-Nonafluorohept-2-en-1-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Nonafluoro-n-butyl)prop-2-enol

009053-1G 009053-5G
101796-214EA 101.54 USD
101796-214 101796-216
1H,1H,2H,3H-Nonafluorohept-2-en-1-ol ≥97%
1H,1H,2H,3H-Nonafluorohept-2-en-1-ol

Hazard: Irritant

Formula: C₇H₅F₉O
Boiling Pt: 78…83 °C (28 mmHg)
MDL Number: MFCD00042340
CAS Number: 72016-05-8

Order Now

Specification Test Results

Boiling point 78-83°C/28m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR