1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)propan-2-ol ≥97%

Supplier: Matrix Scientific

008999-1G 008999-5G
101796-060EA 213.85 USD
101796-060 101796-062
1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)propan-2-ol ≥97%
1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)propan-2-ol

Hazard: Irritant

Formula: C₁₁H₈F₆O
MW: 270.18 g/mol
Boiling Pt: 74…75 °C (3 mmHg)
MDL Number: MFCD04038310
CAS Number: 2386-82-5

Order Now

Specification Test Results

Boiling point 74-75°C/3mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR