1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether ≥98%
Supplier: Matrix Scientific
008996-5G
008996-25G
101796-048EA
130.77
USD
101796-048
101796-050
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether ≥98%
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether
Hazard: Irritant
Formula:
C₄H₂Cl₂F₆O Boiling Pt: 98 °C |
MDL Number:
MFCD03094547 CAS Number: 56860-82-3 |
Specification Test Results
Boiling point | 98°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...