1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane ≥97%

Supplier: Matrix Scientific

008890-1G 008890-5G
101795-728EA 192.31 USD
101795-728 101795-730
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane ≥97%
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane

Hazard: Irritant

Formula: C₅H₄Br₂F₆
Boiling Pt: 92 °C (47 mmHg)
MDL Number: MFCD03095411
CAS Number: 33619-78-2

Order Now

Specification Test Results

Boiling point 92°C/47mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR