3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane

Supplier: Matrix Scientific

002023-1G 002023-5G
101783-068EA 69.23 USD
101783-068 101783-070
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane

Hazard: Irritant

Formula: C₄H₂ClF₆I
Boiling Pt: 124…125 °C
MDL Number: MFCD02093304
CAS Number: 240122-22-9

Order Now

Specification Test Results

Boiling point 124-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR