4-Isopropylbenzyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Chloromethyl)-4-isopropylbenzene

006628-5G 006628-25G
101790-840EA 56.92 USD
101790-840 101790-842
4-Isopropylbenzyl chloride ≥98%
4-Isopropylbenzyl chloride

TCA listed: T
Hazard: Irritant/Lachrymator

Formula: C₁₀H₁₃Cl
MW: 168.67 g/mol
Boiling Pt: 119…120 °C (35 mmHg)
Density: 1.01
MDL Number: MFCD00018885
CAS Number: 2051-18-5
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Boiling point 119-120°C/3
Density 1.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR