Heptafluoro-1-methoxypropane ≥98%
Supplier: Matrix Scientific
Synonyms:
Heptafluoropropyl methyl ether
005898-50G
101789-714EA
235.38
USD
101789-714
Heptafluoro-1-methoxypropane ≥98%
Heptafluoro-1-methoxypropane
TCA listed: T
Hazard: Irritant
Formula:
C₄H₃F₇O MW: 200.06 g/mol Boiling Pt: 34.18 °C Density: 1.409 |
MDL Number:
MFCD01320779 CAS Number: 375-03-1 |
Specification Test Results
Boiling point | 34.18°C |
Density | 1.409 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...