1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥99%
Supplier: Matrix Scientific
Synonyms:
Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate
001198-25G
001198-100G
101781-130EA
58.46
USD
101781-130
101781-132
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥99%
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
TCA listed: T
Hazard: Flammable/Refrigerate
Formula:
C₇H₆F₆O₂ Boiling Pt: 99 °C Density: 1.302 |
MDL Number:
MFCD00040105 CAS Number: 3063-94-3 |
Specification Test Results
Boiling point | 99°C |
Density | 1.302 |
ND20 | 1.331 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...