2,3,4,5,6-Pentafluorobenzhydrol ≥97%
Supplier: Matrix Scientific
Synonyms:
2,3,4,5,6-Pentafluoro-α-phenylbenzenemethanol
001554-1G
001554-5G
101781-962EA
453.85
USD
101781-962
101781-960
2,3,4,5,6-Pentafluorobenzhydrol ≥97%
2,3,4,5,6-Pentafluorobenzhydrol
Hazard: Irritant
Specification Test Results
Boiling point | 115-117°C/1 |
Melting point | 54-57°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...