4-Biphenylmethyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Chloromethyl)biphenyl

056898-1G 056898-5G
101911-644EA 21.54 USD
101911-644 101911-646
4-Biphenylmethyl chloride ≥98%
4-Biphenylmethyl chloride

Hazard: Irritant

Formula: C₁₃H₁₁Cl
Melting Pt: 71…73 °C
MDL Number: MFCD00000917
CAS Number: 1667-11-4

Order Now

Specification Test Results

Melting point 71-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR