1-(4-Isopropylphenyl)-2-thiourea
Supplier: Matrix Scientific
056745-5G
101911-314EA
230.77
USD
101911-314
1-(4-Isopropylphenyl)-2-thiourea
1-(4-Methylphenyl)-2-thiourea
Hazard: Irritant
Formula:
C₁₀H₁₄N₂S Melting Pt: 147…149 °C |
CAS Number:
65259-91-8 |
Specification Test Results
Melting point | 147-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...