tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
, 3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine, N1-BOC protected
055502-500MG
101909-548EA
1109.23
USD
101909-548
tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂F₃IN₂O₂ MW: 412.15 g/mol Melting Pt: 125…127 °C |
MDL Number:
MFCD16622804 CAS Number: 1228183-72-9 |
Specification Test Results
Melting point | 125-127°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...