Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]acetate ≥95%
Supplier: Matrix Scientific
054906-500MG
054906-1G
054906-5G
101909-010EA
181.54
USD
101909-010
101909-012
101909-014
Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]acetate ≥95%
Methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]acetate
Hazard: Irritant
Formula:
C₈H₈N₂O₄S Melting Pt: 91…93 °C |
MDL Number:
MFCD15200563 CAS Number: 224635-20-5 |
Specification Test Results
Melting point | 91-93°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...