4-(2-Pyrimidinyloxy)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Pyrimidin-2-yloxy)benzonitrile

048155-500MG 048155-1G 048155-5G
101883-084EA 181.54 USD
101883-084 101883-086 101883-088
4-(2-Pyrimidinyloxy)benzenecarbonitrile ≥95%
4-(2-Pyrimidinyloxy)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₁H₇N₃O
Melting Pt: 111…113 °C
MDL Number: MFCD01927046
CAS Number: 353259-03-7

Order Now

Specification Test Results

Melting point 111-113°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR