(6-Bromo-1-benzothiophen-2-yl)methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
(6-Bromobenzo[b]thiophen-2-yl)methanol
048103-500MG
048103-1G
101882-810EA
523.08
USD
101882-810
101882-812
(6-Bromo-1-benzothiophen-2-yl)methanol ≥95%
(6-Bromo-1-benzothiophen-2-yl)methanol
Hazard: Irritant
Formula:
C₉H₇BrOS Melting Pt: 107…109 °C |
MDL Number:
MFCD11841039 CAS Number: 374933-76-3 |
Specification Test Results
Melting point | 107-109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...