Methyl-6-piperazinonicotinate ≥95%
Supplier: Matrix Scientific
046631-500MG
046631-1G
101876-342EA
600
USD
101876-342
101876-344
Methyl-6-piperazinonicotinate ≥95%
Methyl-6-piperazinonicotinate
Hazard: Irritant
Formula:
C₁₁H₁₅N₃O₂ Melting Pt: 165…167 °C |
MDL Number:
MFCD00894966 CAS Number: 132144-03-7 |
Specification Test Results
Melting point | 165-167°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...