1-Acetyl-2-methyl-5-indolinesulfonoyl chloride ≥95%

Supplier: Matrix Scientific

046600-500MG 046600-1G 046600-5G
101876-230EA 338.46 USD
101876-230 101876-232 101876-234
1-Acetyl-2-methyl-5-indolinesulfonoyl chloride ≥95%
1-Acetyl-2-methyl-5-indolinesulfonoyl chloride

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃S
Melting Pt: 143…145 °C
MDL Number: MFCD07841689
CAS Number: 841275-78-3

Order Now

Specification Test Results

Melting point 143-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR