6-Chloro-5-iodonicotinic acid ≥95%

Supplier: Matrix Scientific

046180-500MG 046180-1G
101874-554EA 300 USD
101874-554 101874-556
6-Chloro-5-iodonicotinic acid ≥95%
6-Chloro-5-iodonicotinic acid

Hazard: Irritant

Formula: C₆H₃ClINO₂
Melting Pt: 196…198 °C
MDL Number: MFCD11100073
CAS Number: 59782-87-5

Order Now

Specification Test Results

Melting point 196-198°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR