2-(2,4-Difluorophenoxy)ethanethioamide ≥95%

Supplier: Matrix Scientific

046072-500MG 046072-1G
101874-024EA 556.92 USD
101874-024 101874-026
2-(2,4-Difluorophenoxy)ethanethioamide ≥95%
2-(2,4-Difluorophenoxy)ethanethioamide

Hazard: Irritant

Formula: C₈H₇F₂NOS
Melting Pt: 114…115 °C
MDL Number: MFCD09152699
CAS Number: 952182-86-4

Order Now

Specification Test Results

Melting point 114-115°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR