Methyl-3,5-diiodo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

045995-500MG 045995-1G 045995-5G
101873-640EA 278.46 USD
101873-640 101873-642 101873-644
Methyl-3,5-diiodo-1H-indole-2-carboxylate ≥95%
Methyl-3,5-diiodo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₇I₂NO₂
Melting Pt: 209…211 °C
MDL Number: MFCD06200993
CAS Number: 885949-47-3

Order Now

Specification Test Results

Melting point 209-211°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR